Assay Details
| Gene Symbol: LOC101111911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015101737.2 | XP_014957223.2 | 1 - 2 | 973 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101111911| Gene Symbol: LOC101111911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015101737.2 | XP_014957223.2 | 1 - 2 | 973 | 78 | |
Set Membership: |
|