Assay Details
| Gene Symbol: CRHBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009339.1 | NP_001009339.1 | 6 - 7 | 870 | 75 | |
| GenBank mRNA | U51199.1 | - | 6 - 7 | 911 | 75 | |
More Information
Set Membership: |
|

CRHBP
Genomic Map