Assay Details
| Gene Symbol: ZNF131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012096970.3 | XP_011952360.2 | 5 - 6 | 1166 | 131 | |
| XM_012096971.2 | XP_011952361.2 | 5 - 6 | 1277 | 131 | ||
| XM_012096972.3 | XP_011952362.2 | 5 - 6 | 1135 | 131 | ||
| XM_012096974.3 | XP_011952364.2 | 5 - 6 | 1117 | 131 | ||
| XM_027980089.1 | XP_027835890.1 | 2 - 3 | 315 | 131 | ||
More Information
Set Membership: |
|

ZNF131
Genomic Map