Assay Details
| Gene Symbol: NRSN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014464.4 | XP_004014513.1 | 2 - 3 | 241 | 63 | |
| XM_012188984.3 | XP_012044374.1 | 2 - 3 | 310 | 63 | ||
More Information
Set Membership: |
|

NRSN2
Genomic Map