Assay Details
| Gene Symbol: MAP1S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969443.1 | XP_027825244.1 | 1 - 2 | 120 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAP1S| Gene Symbol: MAP1S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969443.1 | XP_027825244.1 | 1 - 2 | 120 | 65 | |
Set Membership: |
|