Assay Details
| Gene Symbol: ZZZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092054.2 | XP_014947540.2 | 8 - 9 | 2436 | 60 | |
| XM_027971360.1 | XP_027827161.1 | 8 - 9 | 2449 | 60 | ||
| XM_027971366.1 | XP_027827167.1 | 6 - 7 | 2548 | 60 | ||
| XM_027971371.1 | XP_027827172.1 | 7 - 8 | 2625 | 60 | ||
More Information
Set Membership: |
|

ZZZ3
Genomic Map