Assay Details
| Gene Symbol: NLRP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008740.4 | XP_004008789.2 | 6 - 7 | 2426 | 58 | |
| XM_027970057.1 | XP_027825858.1 | 4 - 5 | 3421 | 58 | ||
| XM_027970058.1 | XP_027825859.1 | 4 - 5 | 3421 | 58 | ||
| XM_027970059.1 | XP_027825860.1 | 4 - 5 | 3421 | 58 | ||
More Information
Set Membership: |
|

NLRP3
Genomic Map