Assay Details
| Gene Symbol: KCNIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020130.4 | XP_004020179.1 | 4 - 5 | 2228 | 64 | |
| XM_004020133.3 | XP_004020182.1 | 4 - 5 | 2232 | 64 | ||
| XM_012102825.2 | XP_011958215.1 | 4 - 5 | 2232 | 64 | ||
| XM_027960673.1 | XP_027816474.1 | 4 - 5 | 373 | 64 | ||
| XM_027960674.1 | XP_027816475.1 | 4 - 5 | 373 | 64 | ||
| XM_027960675.1 | XP_027816476.1 | 6 - 7 | 568 | 64 | ||
More Information
Set Membership: |
|

KCNIP2
Genomic Map