Assay Details
| Gene Symbol: RHBDF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961228.1 | XP_027817029.1 | 13 - 14 | 1847 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RHBDF1| Gene Symbol: RHBDF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961228.1 | XP_027817029.1 | 13 - 14 | 1847 | 58 | |
Set Membership: |
|