Assay Details
| Gene Symbol: FYB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017027.3 | XP_004017076.2 | 7 - 8 | 1643 | 195 | |
| XM_012097013.3 | XP_011952403.1 | 7 - 8 | 1642 | 195 | ||
| XM_012097014.2 | XP_011952404.1 | 7 - 8 | 1682 | 195 | ||
| XM_012097016.3 | XP_011952406.1 | 7 - 8 | 1858 | 195 | ||
More Information
Set Membership: |
|

FYB1
Genomic Map