Assay Details
| Gene Symbol: HSPA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008795.4 | XP_004008844.1 | 4 - 5 | 655 | 76 | |
| XM_012178480.3 | XP_012033870.1 | 2 - 3 | 990 | 76 | ||
More Information
Set Membership: |
|

HSPA4
Genomic Map