Assay Details
| Gene Symbol: CEP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020791.3 | XP_004020840.1 | 2 - 3 | 281 | 127 | |
| XM_012104150.2 | XP_011959540.1 | 2 - 3 | 281 | 127 | ||
| XM_012104152.3 | XP_011959542.1 | 2 - 3 | 281 | 127 | ||
More Information
Set Membership: |
|

CEP20
Genomic Map