Assay Details
| Gene Symbol: TEAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978605.1 | XP_027834406.1 | 9 - 10 | 1030 | 61 | |
| XM_027978606.1 | XP_027834407.1 | 9 - 10 | 816 | 61 | ||
| XM_027978607.1 | XP_027834408.1 | 7 - 8 | 1565 | 61 | ||
More Information
Set Membership: |
|

TEAD2
Genomic Map