Assay Details
| Gene Symbol: RFXANK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008404.4 | XP_004008453.1 | 4 - 5 | 598 | 57 | |
| XM_012177671.3 | XP_012033061.1 | 5 - 6 | 577 | 57 | ||
| XM_012177674.3 | XP_012033064.1 | 4 - 5 | 603 | 57 | ||
| XM_012177675.3 | XP_012033065.1 | 4 - 5 | 595 | 57 | ||
| XM_027969353.1 | XP_027825154.1 | 5 - 6 | 574 | 57 | ||
More Information
Set Membership: |
|

RFXANK
Genomic Map