Assay Details
| Gene Symbol: PPIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004617.4 | XP_004004666.2 | 7 - 8 | 568 | 82 | |
| XM_012134405.3 | XP_011989795.2 | 7 - 8 | 625 | 82 | ||
| XM_012134426.3 | XP_011989816.2 | 8 - 9 | 675 | 82 | ||
| XM_012134434.3 | XP_011989824.2 | 8 - 9 | 697 | 82 | ||
| XM_012134444.3 | XP_011989834.2 | 8 - 9 | 618 | 82 | ||
| XM_012134460.3 | XP_011989850.2 | 7 - 8 | 560 | 82 | ||
| XM_012134469.2 | XP_011989859.2 | 8 - 9 | 556 | 82 | ||
| XM_012134474.3 | XP_011989864.2 | 7 - 8 | 546 | 82 | ||
| XM_012134484.3 | XP_011989874.2 | 8 - 9 | 730 | 82 | ||
| XM_012134507.2 | XP_011989897.2 | 7 - 8 | 503 | 82 | ||
| XM_012134516.2 | XP_011989906.2 | 7 - 8 | 495 | 82 | ||
More Information
Set Membership: |
|

PPIG
Genomic Map