Assay Details
| Gene Symbol: KLF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965127.1 | XP_027820928.1 | 4 - 5 | 1129 | 63 | |
| XM_027965128.1 | XP_027820929.1 | 4 - 5 | 1126 | 63 | ||
More Information
Set Membership: |
|

KLF7
Genomic Map