Assay Details
| Gene Symbol: LOC101111980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974883.1 | XP_027830684.1 | 35 - 36 | 5491 | 73 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101111980| Gene Symbol: LOC101111980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974883.1 | XP_027830684.1 | 35 - 36 | 5491 | 73 | |