Assay Details
| Gene Symbol: CCND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127289.1 | NP_001120761.1 | 4 - 5 | 711 | 67 | |
| XM_027958330.1 | XP_027814131.1 | 4 - 5 | 713 | 67 | ||
| XM_027958331.1 | XP_027814132.1 | 3 - 4 | 500 | 67 | ||
| GenBank mRNA | EU626222.1 | - | 4 - 5 | 711 | 67 | |
More Information
Set Membership: |
|

CCND3
Genomic Map