Assay Details
| Gene Symbol: LOC101108898 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966456.1 | XP_027822257.1 | 1 - 2 | 122 | 64 | |
| Gene Symbol: LOC114113882 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966457.1 | XP_027822258.1 | 1 - 2 | 168 | 64 | |
More Information
Set Membership: |
|

LOC101108898
Genomic Map