Assay Details
| Gene Symbol: COX15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020102.4 | XP_004020151.1 | 6 - 7 | 923 | 53 | |
| XM_027960444.1 | XP_027816245.1 | 5 - 6 | 651 | 53 | ||
| GenBank mRNA | DQ152943.1 | - | 2 - 3 | 113 | 53 | |
More Information
Set Membership: |
|

COX15
Genomic Map