Assay Details
| Gene Symbol: IL17RD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958261.1 | XP_027814062.1 | 5 - 6 | 905 | 61 | |
| XM_027958262.1 | XP_027814063.1 | 6 - 7 | 634 | 61 | ||
More Information
Set Membership: |
|

IL17RD
Genomic Map