Assay Details
| Gene Symbol: METTL23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013104.4 | XP_004013153.1 | 2 - 3 | 494 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

METTL23| Gene Symbol: METTL23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013104.4 | XP_004013153.1 | 2 - 3 | 494 | 61 | |
Set Membership: |
|