Assay Details
| Gene Symbol: LOC101111419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961544.1 | XP_027817345.1 | 1 - 2 | 1995 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101111419| Gene Symbol: LOC101111419 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961544.1 | XP_027817345.1 | 1 - 2 | 1995 | 62 | |
Set Membership: |
|