Assay Details
| Gene Symbol: NCKAP5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006373.4 | XP_004006422.3 | 8 - 9 | 3344 | 68 | |
| XM_027967354.1 | XP_027823155.1 | 9 - 10 | 3508 | 68 | ||
| XM_027967355.1 | XP_027823156.1 | 9 - 10 | 3478 | 68 | ||
| XM_027967356.1 | XP_027823157.1 | 9 - 10 | 3441 | 68 | ||
| XM_027967357.1 | XP_027823158.1 | 9 - 10 | 3411 | 68 | ||
| XM_027967358.1 | XP_027823159.1 | 8 - 9 | 3279 | 68 | ||
More Information
Set Membership: |
|

NCKAP5L
Genomic Map