Assay Details
| Gene Symbol: PRKAA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112816.1 | NP_001106287.1 | 3 - 4 | 332 | 68 | |
| XM_027965800.1 | XP_027821601.1 | 3 - 4 | 376 | 68 | ||
| GenBank mRNA | EU131097.1 | - | 3 - 4 | 332 | 68 | |
More Information
Set Membership: |
|

PRKAA2
Genomic Map