Assay Details
| Gene Symbol: PKM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010279.4 | XP_004010328.1 | 3 - 4 | 555 | 71 | |
| XM_012180891.3 | XP_012036281.1 | 3 - 4 | 467 | 71 | ||
More Information
Set Membership: |
|

PKM
Genomic Map