Assay Details
Gene Symbol: LOC101113131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963039.1 | XP_027818840.1 | 1 - 2 | 1171 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
