Assay Details
| Gene Symbol: KLF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096439.2 | XP_014951925.1 | 4 - 5 | 1048 | 76 | |
| XM_027970929.1 | XP_027826730.1 | 3 - 4 | 4708 | 76 | ||
More Information
Set Membership: |
|

KLF3
Genomic Map