Assay Details
| Gene Symbol: TMEM165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971006.1 | XP_027826807.1 | 2 - 3 | 709 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM165| Gene Symbol: TMEM165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971006.1 | XP_027826807.1 | 2 - 3 | 709 | 63 | |
Set Membership: |
|