Assay Details
| Gene Symbol: LOC101114198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011124.4 | XP_004011173.2 | 10 - 11 | 1976 | 93 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101114198| Gene Symbol: LOC101114198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011124.4 | XP_004011173.2 | 10 - 11 | 1976 | 93 | |