Assay Details
| Gene Symbol: PPIL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011404.4 | XP_004011453.2 | 11 - 12 | 1121 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPIL4| Gene Symbol: PPIL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011404.4 | XP_004011453.2 | 11 - 12 | 1121 | 69 | |
Set Membership: |
|