Assay Details
| Gene Symbol: LOC101122744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979022.1 | XP_027834823.1 | 12 - 13 | 3031 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101122744| Gene Symbol: LOC101122744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979022.1 | XP_027834823.1 | 12 - 13 | 3031 | 73 | |
Set Membership: |
|