Assay Details
| Gene Symbol: DNAAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010592.3 | XP_004010641.1 | 3 - 4 | 464 | 128 | |
| XM_004010593.3 | XP_004010642.1 | 3 - 4 | 464 | 128 | ||
| XM_015097028.2 | XP_014952514.1 | 3 - 4 | 464 | 128 | ||
| XM_015097029.2 | XP_014952515.1 | 3 - 4 | 461 | 128 | ||
| XM_015097030.2 | XP_014952516.1 | 3 - 4 | 396 | 128 | ||
| XM_027971807.1 | XP_027827608.1 | 3 - 4 | 464 | 128 | ||
More Information
Set Membership: |
|

DNAAF4
Genomic Map