Assay Details
| Gene Symbol: KCNMB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006463.4 | XP_004006512.1 | 1 - 2 | 619 | 105 | |
| GenBank mRNA | JF499673.1 | - | 1 - 2 | 258 | 105 | |
More Information
Set Membership: |
|

KCNMB4
Genomic Map