Assay Details
| Gene Symbol: LOC114116372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973017.1 | XP_027828818.1 | 10 - 11 | 1398 | 90 | |
| XM_027973018.1 | XP_027828819.1 | 10 - 11 | 1401 | 90 | ||
More Information
Set Membership: |
|

LOC114116372
Genomic Map