Assay Details
| Gene Symbol: MRE11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012095281.2 | XP_011950671.2 | 16 - 17 | 2077 | 71 | |
| XM_012095282.3 | XP_011950672.2 | 16 - 17 | 2041 | 71 | ||
| XM_012095283.2 | XP_011950673.2 | 14 - 15 | 1703 | 71 | ||
| XM_012095284.2 | XP_011950674.2 | 13 - 14 | 1631 | 71 | ||
More Information
Set Membership: |
|

MRE11
Genomic Map