Assay Details
| Gene Symbol: LOC101121595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968820.1 | XP_027824621.1 | 1 - 2 | 1185 | 101 | |
| XM_027968821.1 | XP_027824622.1 | 1 - 2 | 1198 | 101 | ||
| XM_027968822.1 | XP_027824623.1 | 1 - 2 | 1187 | 101 | ||
| XM_027968825.1 | XP_027824626.1 | 1 - 2 | 299 | 101 | ||
| XM_027968826.1 | XP_027824627.1 | 1 - 2 | 1182 | 101 | ||
| XM_027968827.1 | XP_027824628.1 | 1 - 2 | 286 | 101 | ||
| XM_027968828.1 | XP_027824629.1 | 1 - 2 | 278 | 101 | ||
More Information
Set Membership: |
|

LOC101121595
Genomic Map