Assay Details
| Gene Symbol: ZNF608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969978.1 | XP_027825779.1 | 3 - 4 | 2776 | 62 | |
| XM_027969979.1 | XP_027825780.1 | 3 - 4 | 1778 | 62 | ||
| XM_027969980.1 | XP_027825781.1 | 3 - 4 | 2776 | 62 | ||
| XM_027969981.1 | XP_027825782.1 | 2 - 3 | 669 | 62 | ||
More Information
Set Membership: |
|

ZNF608
Genomic Map