Assay Details
| Gene Symbol: UNC13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971817.1 | XP_027827618.1 | 2 - 3 | 3019 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UNC13C| Gene Symbol: UNC13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971817.1 | XP_027827618.1 | 2 - 3 | 3019 | 64 | |
Set Membership: |
|