Assay Details
| Gene Symbol: LOC101118793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972290.1 | XP_027828091.1 | 18 - 19 | 2458 | 90 | |
| XM_027972291.1 | XP_027828092.1 | 17 - 18 | 2437 | 90 | ||
| XM_027972292.1 | XP_027828093.1 | 11 - 12 | 1785 | 90 | ||
More Information
Set Membership: |
|

LOC101118793
Genomic Map