Assay Details
| Gene Symbol: LOC101118793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972290.1 | XP_027828091.1 | 5 - 6 | 512 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101118793| Gene Symbol: LOC101118793 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972290.1 | XP_027828091.1 | 5 - 6 | 512 | 90 | |
Set Membership: |
|