Assay Details
| Gene Symbol: LOC554335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974354.1 | XP_027830155.1 | 9 - 10 | 1497 | 70 | |
| XM_027974355.1 | XP_027830156.1 | 3 - 4 | 463 | 70 | ||
| GenBank mRNA | AJ920033.1 | - | 2 - 3 | 348 | 70 | |
More Information
Set Membership: |
|

LOC554335
Genomic Map