Assay Details
| Gene Symbol: FOXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972113.1 | XP_027827914.1 | 4 - 5 | 990 | 78 | |
| XM_027972114.1 | XP_027827915.1 | 4 - 5 | 2857 | 78 | ||
| XM_027972115.1 | XP_027827916.1 | 3 - 4 | 756 | 78 | ||
| XM_027972116.1 | XP_027827917.1 | 3 - 4 | 1004 | 78 | ||
| XM_027972117.1 | XP_027827918.1 | 4 - 5 | 990 | 78 | ||
| XM_027972118.1 | XP_027827919.1 | 4 - 5 | 990 | 78 | ||
More Information
Set Membership: |
|

FOXN3
Genomic Map