Assay Details
| Gene Symbol: TCTEX1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012096392.3 | XP_011951782.2 | 4 - 5 | 594 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TCTEX1D2| Gene Symbol: TCTEX1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012096392.3 | XP_011951782.2 | 4 - 5 | 594 | 74 | |
Set Membership: |
|