Assay Details
| Gene Symbol: SLC6A17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012178160.3 | XP_012033550.1 | 9 - 10 | 1973 | 61 | |
| XM_012178162.3 | XP_012033552.1 | 9 - 10 | 2261 | 61 | ||
| XM_027972874.1 | XP_027828675.1 | 8 - 9 | 1492 | 61 | ||
More Information
Set Membership: |
|

SLC6A17
Genomic Map