Assay Details
Gene Symbol: MGARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017244.3 | XP_004017293.2 | 2 - 3 | 219 | 71 | |
XM_027956267.1 | XP_027812068.1 | 2 - 3 | 151 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
