Assay Details
| Gene Symbol: VGLL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978987.1 | XP_027834788.1 | 2 - 3 | 475 | 88 | |
| XM_027978988.1 | XP_027834789.1 | 2 - 3 | 472 | 88 | ||
| XM_027978989.1 | XP_027834790.1 | 2 - 3 | 307 | 88 | ||
More Information
Set Membership: |
|

VGLL3
Genomic Map