Assay Details
| Gene Symbol: FGF14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015097958.2 | XP_014953444.2 | 4 - 5 | 613 | 64 | |
| XM_027973760.1 | XP_027829561.1 | 4 - 5 | 1482 | 64 | ||
More Information
Set Membership: |
|

FGF14
Genomic Map