Assay Details
| Gene Symbol: OTUD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956254.1 | XP_027812055.1 | 17 - 18 | 1732 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

OTUD4| Gene Symbol: OTUD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956254.1 | XP_027812055.1 | 17 - 18 | 1732 | 60 | |
Set Membership: |
|