Assay Details
| Gene Symbol: EPRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013603.4 | XP_004013652.2 | 20 - 21 | 3153 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPRS1| Gene Symbol: EPRS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013603.4 | XP_004013652.2 | 20 - 21 | 3153 | 85 | |
Set Membership: |
|